Learn More
ACROBiosystems Human FAP Protein, Fc Tag
Catalog No. 502019651
Encompass_Preferred
Recombinant Protein;100UG;Human FAP, Fc Tag (FAP-H5263) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # Q12884-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 111.5 kDa. The protein migrates as 110-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FAP, Fc Tag (Cat. No. FAP-H5263) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-FAP (FAP5) Antibody, Mouse IgG2a with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Monoclonal Anti-FAP (FAP5) Antibody, Mouse IgG2a at 1 μg/mL (100 μL/well) can bind Human FAP, Fc Tag (Cat. No. FAP-H5263) with a linear range of 1-8 ng/mL (Routinely tested).
Catalog No. 50-201-9651 Supplier ACROBiosystems Supplier No. FAPH5263100UG
Recombinant Protein;100UG;Human FAP, Fc Tag (FAP-H5263) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # Q12884-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 111.5 kDa. The protein migrates as 110-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FAP, Fc Tag (Cat. No. FAP-H5263) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-FAP (FAP5) Antibody, Mouse IgG2a with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Monoclonal Anti-FAP (FAP5) Antibody, Mouse IgG2a at 1 μg/mL (100 μL/well) can bind Human FAP, Fc Tag (Cat. No. FAP-H5263) with a linear range of 1-8 ng/mL (Routinely tested).