Learn More
ACROBiosystems Human FAP Protein, His Tag
Supplier:  ACROBiosystems FAPH5244100UG
Encompass_Preferred
Recombinant Protein;100UG;Human FAP, His Tag (FAP-H5244) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # Q12884-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 86.9 kDa.
Catalog No. 50-201-9653