Learn More
ACROBiosystems Human BAFF / TNFSF13B / CD257 Protein, Fc Tag, active trimer (MALS verified)
Catalog No. 502019691
Encompass_Preferred
Recombinant Protein;100UG;Human BAFF, Fc Tag, active trimer (BAF-H5261) is expressed from human 293 cells (HEK293). It contains AA Glu 140 - Leu 285 (Accession # Q9Y275-1) trimer Design.;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 76.7 kDa. The protein migrates as 120-140 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BCA-H5253) at 1 μg/mL (100 μL/well) can bind Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) with a linear range of 0.2-6 ng/mL (QC tested).;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) at 1 μg/mL (100 μL/well) can bind Biotinylated Human TACI, Fc,Avitag (Cat. No. TAI-H82F6) with a linear range of 0.002-0.05 μg/mL (Routinely tested).;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) on CM5 Chip can bind Human TACI/TNFRSF13B, Fc Tag (Cat. No. TAI-H5256) with an affinity constant of 0.658
Catalog No. 50-201-9691 Supplier ACROBiosystems Supplier No. BAFH5261100UG
Recombinant Protein;100UG;Human BAFF, Fc Tag, active trimer (BAF-H5261) is expressed from human 293 cells (HEK293). It contains AA Glu 140 - Leu 285 (Accession # Q9Y275-1) trimer Design.;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 76.7 kDa. The protein migrates as 120-140 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BCA-H5253) at 1 μg/mL (100 μL/well) can bind Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) with a linear range of 0.2-6 ng/mL (QC tested).;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) at 1 μg/mL (100 μL/well) can bind Biotinylated Human TACI, Fc,Avitag (Cat. No. TAI-H82F6) with a linear range of 0.002-0.05 μg/mL (Routinely tested).;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) on CM5 Chip can bind Human TACI/TNFRSF13B, Fc Tag (Cat. No. TAI-H5256) with an affinity constant of 0.658