Learn More
ACROBiosystems Human Siglec-3 / CD33 Protein, Fc Tag
Catalog No. 502019680
Encompass_Preferred
Recombinant Protein;100UG;Human Siglec-3, Fc Tag (CD3-H5257) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 64-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3 Antibody at 1 μg/mL (100 μL/well) can bind Human Siglec-3, Fc Tag (Cat. No. CD3-H5257) with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-201-9680 Supplier ACROBiosystems Supplier No. CD3H5257100UG
Recombinant Protein;100UG;Human Siglec-3, Fc Tag (CD3-H5257) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 64-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3 Antibody at 1 μg/mL (100 μL/well) can bind Human Siglec-3, Fc Tag (Cat. No. CD3-H5257) with a linear range of 0.2-2 ng/mL (QC tested).