Learn More
ACROBiosystems Human CD8 alpha&beta (CD8A&CD8B) Heterodimer Protein, His Tag&Tag Free (MALS verified)
Supplier:  ACROBiosystems CDAH52W5100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD8A&CD8B Heterodimer Protein, His Tag&Tag Free (CDA-H52W5) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Asp 182 (CD8A) & Leu 22 - Pro 170 (CD8B) (Accession # P01732-1 (CD8A) & P10966-1 (CD8B) ).;Human CD8A&CD8B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of CD8 alpha and CD8 beta, has a calculated MW of 19.5 kDa (CD8 alpha) and 22.1 kDa (CD8 beta). Subunit CD8 alpha is fused with a polyhistidine tag at the C-terminus and subunit CD8 beta contains no tag. As a result of glycosylation, the heterodimer protein migrates as 30-32 kDa and 32-34 kDa under reducing (R) condition, and 30-33 kDa and 60-65 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD8A&CD8B Heterodimer Protein, His Tag&Tag Free (Cat. No. CDA-H52W5) at 1 μg/mL (100 μL/well) can bind Anti-CD8 alpha Antibody, Mouse IgG2a (clone: OKT8) with a linear range of 0.2-5 ng/mL (QC tested).;Immobilized Anti-CD8 alpha Antibody, Mouse IgG2a
Catalog No. 50-201-9730