Learn More
ACROBiosystems Human Semaphorin 4D / SEMA4D / CD100 Protein, Fc Tag
Supplier:  ACROBiosystems CD0H5257100UG
Encompass_Preferred
Recombinant Protein;100UG;Human SEMA4D, Fc Tag (CD0-H5257) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Arg 734 (Accession # AAH54500).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 105.4 kDa. The protein migrates as 125-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9447