Learn More
ACROBiosystems Human Mucin-1 / MUC-1 (24-380) Protein, Fc Tag
Supplier:  ACROBiosystems MU1H5254100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Mucin-1 (24-380), Fc Tag (MU1-H5254) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 380 (Accession # P15941-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.2 kDa.
Catalog No. 50-201-9756