Learn More
ACROBiosystems Human IL-31 Protein, His Tag
Catalog No. 502019760
Encompass_Preferred
Recombinant Protein;100UG;Human IL-31, His Tag (IL1-H5247) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Thr 164 (Accession # Q6EBC2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.7 kDa. The protein migrates as 23-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-31, His Tag (Cat. No. IL1-H5247) at 5 μg/mL (100 μL/well) can bind Human IL-31 RA, Fc Tag (Cat. No. ILA-H5256) with a linear range of 0.02-0.625 μg/mL (QC tested).
Catalog No. 50-201-9760 Supplier ACROBiosystems Supplier No. IL1H5247100UG
Recombinant Protein;100UG;Human IL-31, His Tag (IL1-H5247) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Thr 164 (Accession # Q6EBC2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.7 kDa. The protein migrates as 23-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-31, His Tag (Cat. No. IL1-H5247) at 5 μg/mL (100 μL/well) can bind Human IL-31 RA, Fc Tag (Cat. No. ILA-H5256) with a linear range of 0.02-0.625 μg/mL (QC tested).