Learn More
ACROBiosystems Human PSMA / FOLH1 Protein, Fc Tag
Catalog No. 502019787
Encompass_Preferred
Recombinant Protein;100UG;Human PSMA, Fc Tag (PSA-H5264) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1 ).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 106.0 kDa. The protein migrates as 110-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PSMA, Fc Tag (Cat. No. PSA-H5264) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human PSMA Antibody, Human IgG1 with a linear range of 0.1-2 ng/mL (QC tested).
Catalog No. 50-201-9787 Supplier ACROBiosystems Supplier No. PSAH5264100UG
Recombinant Protein;100UG;Human PSMA, Fc Tag (PSA-H5264) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1 ).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 106.0 kDa. The protein migrates as 110-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PSMA, Fc Tag (Cat. No. PSA-H5264) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human PSMA Antibody, Human IgG1 with a linear range of 0.1-2 ng/mL (QC tested).