Learn More
ACROBiosystems Human GUCY2C / Guanylyl cyclase C Protein, His Tag
Supplier: ACROBiosystems GUCH52H5100UG
Recombinant Protein;100UG;Human GUCY2C, His Tag (GUC-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 55-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GUCY2C, His Tag (Cat. No. GUC-H52H5) at 5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (QC tested).;2e5 of anti-GUCY2C CAR-293 cells were stained with 100 µL of 1 µg/mL Human GUCY2C, His Tag (Cat. No. GUC-H52H5) and negative control protein respectively, washed and then followed by PE anti-His antibody and analyzed with FACS (Routinely tested).