Learn More
ACROBiosystems Human LAG-3 / CD223 Protein, Mouse IgG2a Fc Tag
Supplier: ACROBiosystems LA3H52AA100UG
Recombinant Protein;100UG;Human LAG-3, Mouse IgG2a Fc Tag (LA3-H52Aa) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 73.3 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Mouse IgG2a Fc Tag (Cat. No. LA3-H52Aa) at 2 μg/mL (100 μL/well) can bind Anti-Human LAG3 MAb, Human IgG1 with a linear range of 1-10 ng/mL (QC tested).;Immobilized Human LAG-3, Mouse IgG2a Fc Tag (Cat. No. LA3-H52Aa) at 10 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque FGL1, Fc Tag (Cat. No. FG1-C5269) with a linear range of 0.01-0.156 μg/mL (Routinely tested).;Flow Cytometry assay shows that Human LAG3, Mouse IgG2a Fc Tag (Cat. No. LA3-H52Aa) can bind to daudi cell surface. The concentration of LAG3 used is 0.3 μg/mL (Routinely tested).;FACS analysis shows that the binding Human LAG3