Learn More
ACROBiosystems Human CD99 / MIC2 Protein, His Tag
Supplier:  ACROBiosystems CD9H5221100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession # AAH02584).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 10.9 kDa. The protein migrates as 25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9874