Learn More
ACROBiosystems Human ILDR2 Protein, Fc Tag
Catalog No. 502019459
Encompass_Preferred
Recombinant Protein;100UG;Human ILDR2, Fc Tag (IL2-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Glu 186 (Accession # Q71H61-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.1 kDa.
Catalog No. 50-201-9459 Supplier ACROBiosystems Supplier No. IL2H5259100UG
Recombinant Protein;100UG;Human ILDR2, Fc Tag (IL2-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Glu 186 (Accession # Q71H61-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.1 kDa.