Learn More
ACROBiosystems Human Frizzled-10 / FZD10 Protein, His Tag
Catalog No. 502019464
Encompass_Preferred
Recombinant Protein;100UG;Human Frizzled-10, His Tag (FRD-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Gly 161 (Accession # Q9ULW2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.9 kDa.
Catalog No. 50-201-9464 Supplier ACROBiosystems Supplier No. FRDH52H3100UG
Recombinant Protein;100UG;Human Frizzled-10, His Tag (FRD-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Gly 161 (Accession # Q9ULW2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.9 kDa.