Learn More
ACROBiosystems Human RNF43 Protein, Fc Tag
Catalog No. 502019434
Encompass_Preferred
Recombinant Protein;100UG;Human RNF43, Fc Tag (RN3-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 44 - Tyr 197 (Accession # Q68DV7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.8 kDa.
Catalog No. 50-201-9434 Supplier ACROBiosystems Supplier No. RN3H5256100UG
Recombinant Protein;100UG;Human RNF43, Fc Tag (RN3-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 44 - Tyr 197 (Accession # Q68DV7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.8 kDa.