Learn More
ACROBiosystems Mouse PD-L1 / B7-H1 Protein, Mouse IgG2a Fc Tag, low endotoxin
Supplier:  ACROBiosystems PD1M52A2100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (PD1-M52A2) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # NP_068693).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 51.8 kDa. The protein migrates as 65-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-1, Fc Tag (Cat. No. CD0-M5259) at 5 μg/mL (100 μL/well) can bind Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PD1-M52A2) with a linear range of 0.16-2.5 μg/mL (QC tested).;Immobilized Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PD1-M52A2) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.313-5 μg/mL (Routinely tested).
Catalog No. 50-201-9833