Learn More
ACROBiosystems Mouse GITR Ligand / TNFSF18 Protein, Fc Tag
Catalog No. 502019848
Encompass_Preferred
Recombinant Protein;100UG;Mouse GITR Ligand, Fc Tag (GIL-M526x) is expressed from human 293 cells (HEK293). It contains AA Thr 47 - Ser 173 (Accession # AAO89011).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 41.4 kDa. As a result of glycosylation, the protein migrates as 45 kDa under reducing (R) condition, and 80-100 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse GITR, His Tag (Cat. No. GIR-M5220) at 5 μg/mL (100 μL/well) can bind Mouse GITR Ligand, Fc Tag (Cat. No. GIL-M526x) with a linear range of 5-156 ng/mL (QC tested).
Catalog No. 50-201-9848 Supplier ACROBiosystems Supplier No. GILM526X100UG
Recombinant Protein;100UG;Mouse GITR Ligand, Fc Tag (GIL-M526x) is expressed from human 293 cells (HEK293). It contains AA Thr 47 - Ser 173 (Accession # AAO89011).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 41.4 kDa. As a result of glycosylation, the protein migrates as 45 kDa under reducing (R) condition, and 80-100 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse GITR, His Tag (Cat. No. GIR-M5220) at 5 μg/mL (100 μL/well) can bind Mouse GITR Ligand, Fc Tag (Cat. No. GIL-M526x) with a linear range of 5-156 ng/mL (QC tested).