Learn More
ACROBiosystems Mouse NOTCH1 Protein, His Tag
Catalog No. 502019622
Encompass_Preferred
Recombinant Protein;100UG;Mouse NOTCH1, His Tag (NO1-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Gln 526 (Accession # Q01705-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.4 kDa. The protein migrates as 58-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DLL4, Fc Tag (Cat. No. DL4-H5259) at 5 μg/mL (100 μL/well) can bind Mouse NOTCH1, His Tag (Cat. No. NO1-M52H8) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-201-9622 Supplier ACROBiosystems Supplier No. NO1M52H8100UG
Recombinant Protein;100UG;Mouse NOTCH1, His Tag (NO1-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Gln 526 (Accession # Q01705-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.4 kDa. The protein migrates as 58-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DLL4, Fc Tag (Cat. No. DL4-H5259) at 5 μg/mL (100 μL/well) can bind Mouse NOTCH1, His Tag (Cat. No. NO1-M52H8) with a linear range of 0.02-0.313 μg/mL (QC tested).