Learn More
ACROBiosystems Mouse EpCAM / TROP1 Protein, His Tag
Catalog No. 502019620
Encompass_Preferred
Recombinant Protein;100UG;Mouse EpCAM, His Tag (EPM-M5227) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Thr 266 (Accession # AAH05618.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.5 kDa. The protein migrates as 33-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9620 Supplier ACROBiosystems Supplier No. EPMM5227100UG
Recombinant Protein;100UG;Mouse EpCAM, His Tag (EPM-M5227) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Thr 266 (Accession # AAH05618.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.5 kDa. The protein migrates as 33-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.