Learn More
ACROBiosystems Mouse GUCY2C / Guanylyl cyclase C Protein, Fc Tag
Catalog No. 502019641
Encompass_Preferred
Recombinant Protein;100UG;Mouse GUCY2C, Fc Tag (GUC-M5254) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Met 433 (Accession # Q3UWA6-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9641 Supplier ACROBiosystems Supplier No. GUCM5254100UG
Recombinant Protein;100UG;Mouse GUCY2C, Fc Tag (GUC-M5254) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Met 433 (Accession # Q3UWA6-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.