Learn More
ACROBiosystems SARS Nucleocapsid protein, His Tag
Catalog No. 502019397
Encompass_Preferred
Recombinant Protein;100UG;SARS Nucleocapsid protein, His Tag (NUN-S5229) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 422 (Accession # NP_828858.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.7 kDa. The protein migrates as 30-33 kDa and 55-60 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized SARS Nucleocapsid protein, His Tag (Cat. No. NUN-S5229) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41) with a linear range of 0.1-2 ng/mL (QC tested).
Catalog No. 50-201-9397 Supplier ACROBiosystems Supplier No. NUNS5229100UG
Recombinant Protein;100UG;SARS Nucleocapsid protein, His Tag (NUN-S5229) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 422 (Accession # NP_828858.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.7 kDa. The protein migrates as 30-33 kDa and 55-60 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized SARS Nucleocapsid protein, His Tag (Cat. No. NUN-S5229) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41) with a linear range of 0.1-2 ng/mL (QC tested).