Learn More
ACROBiosystems ActiveMax Human CCL19 / MIP-3 beta Protein, Tag Free
Supplier:  ACROBiosystems CC9H52151MG
Encompass_Preferred
Recombinant Protein;1MG;ActiveMax Human CCL19 / MIP-3 beta, Tag Free (CC9-H5215) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Ser 98 (Accession # Q99731-1).;This protein carries no "tag". The protein has a calculated MW of 8.8 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9774