Learn More
ACROBiosystems Cynomolgus FOLR2 Protein, His Tag
Supplier:  ACROBiosystems FO2C52H81MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus FOLR2, His Tag (FO2-C52H8) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - His 245 (Accession # A0A2K5U027-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 33-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9492