Learn More
ACROBiosystems Cynomolgus IL-31 Protein, His Tag
Catalog No. 502019544
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus IL-31, His Tag (IL1-C5245) is expressed from human 293 cells (HEK293). It contains AA Leu 27 - Thr 163 (Accession # G7PJ78-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.3 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9544 Supplier ACROBiosystems Supplier No. IL1C52451MG
Recombinant Protein;1MG;Cynomolgus IL-31, His Tag (IL1-C5245) is expressed from human 293 cells (HEK293). It contains AA Leu 27 - Thr 163 (Accession # G7PJ78-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.3 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.