Learn More
ACROBiosystems Cynomolgus Latent TGF-beta 1 (C33S) Protein, His Tag
Supplier:  ACROBiosystems TG1C52431MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus Latent TGF-Beta 1 (C33S), His Tag (TG1-C5243) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (C33S) (Accession # A0A2K5TJB2-1 (C33S)).;This protein carries a polyhistidine tag at the N-terminus. The precursor proprotein is partially cleaved by FURIN to form mature Transforming growth factor beta-1 (TGF-beta-1) and Latency-associated peptide (LAP) chains with calculated MW of 12.8 kDa and 30.4 kDa respectively. The protein migrates as 50 kDa (precursor proprotein), 37-45 kDa (LAP) and 13 kDa (TGFB1) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Latent TGF-Beta 1 (C33S), His Tag (Cat. No. TG1-C5243) at 2 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT6-H82E4) with a linear range of 0.6-20 ng/mL (QC tested).;Immobilized Cynomolgus Latent TGF-Beta 1 (C33S), His Tag (Cat. No. TG1-C5243) at 2 μg/mL (100 μL/well) can bind Biot
Catalog No. 50-201-9550