Learn More
ACROBiosystems Cynomolgus GUCY2C / Guanylyl cyclase C Protein, Fc Tag
Supplier:  ACROBiosystems GUCC52551MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus GUCY2C, Fc Tag (GUC-C5255) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # XP_005570270.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.6 kDa. The protein migrates as 95-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus GUCY2C, Fc Tag (Cat. No. GUC-C5255) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 (Cat. No. GUC-M78) with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-201-9642