Learn More
ACROBiosystems Cynomolgus GUCY2C / Guanylyl cyclase C Protein, His Tag
Catalog No. 502019644
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus GUCY2C, His Tag (GUC-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # XP_005570270.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.0 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus GUCY2C, His Tag (Cat. No. GUC-C52H6) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 with a linear range of 0.3-6 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Cynomolgus GUCY2C, His Tag (Cat. No. GUC-C52H6) with a linear range of 3-25 ng/mL (Routinely tested).
Catalog No. 50-201-9644 Supplier ACROBiosystems Supplier No. GUCC52H61MG
Recombinant Protein;1MG;Cynomolgus GUCY2C, His Tag (GUC-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # XP_005570270.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.0 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus GUCY2C, His Tag (Cat. No. GUC-C52H6) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 with a linear range of 0.3-6 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Cynomolgus GUCY2C, His Tag (Cat. No. GUC-C52H6) with a linear range of 3-25 ng/mL (Routinely tested).