Learn More
ACROBiosystems Cynomolgus IL-33 Protein, His Tag
Catalog No. 502019667
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus IL-33, His Tag (IL3-C5245) is expressed from human 293 cells (HEK293). It contains AA His 109 - Ile 270 (Accession # A0A2K5W3I1-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus IL-33, His Tag (Cat. No. IL3-C5245) at 5 μg/mL (100 μL/well) can bind Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) with a linear range of 2-78 ng/mL (QC tested).
Catalog No. 50-201-9667 Supplier ACROBiosystems Supplier No. IL3C52451MG
Recombinant Protein;1MG;Cynomolgus IL-33, His Tag (IL3-C5245) is expressed from human 293 cells (HEK293). It contains AA His 109 - Ile 270 (Accession # A0A2K5W3I1-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus IL-33, His Tag (Cat. No. IL3-C5245) at 5 μg/mL (100 μL/well) can bind Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) with a linear range of 2-78 ng/mL (QC tested).