Learn More
ACROBiosystems Cynomolgus IL-23 alpha & Mouse IL-12 beta Heterodimer Protein, His Tag&Tag Free
Catalog No. 502019790
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus IL-23A & Mouse IL-12B Heterodimer Protein, His Tag&Tag Free (ILB-CM52W8) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Pro 189 (IL23A) & Met 23 - Ser 335 (IL12B) (Accession # A0A2K5TLV4-1 (IL23A) & P43432-1 (IL12B)).;Cynomolgus IL-23A & Mouse IL-12B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of IL-23 alpha and IL-12 beta, has a calculated MW of 20.3 kDa (IL-23 alpha) and 35.8 kDa (IL-12 beta). Subunit IL-23 alpha is fused with a polyhistidine tag at the N-terminus and subunit IL-12 beta contains no tag. The reducing (R) protein migrates as 20-23 kDa (IL-23 alpha) and 40-42 kDa and 43-46 kDa (IL-12 beta) respectively due to glycosylation.;Immobilized Biotinylated Human IL-23 R, Fc,Avitag (Cat. No. ILR-H82F3) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Cynomolgus IL-23A & Mouse IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-CM52W8) with a li
Catalog No. 50-201-9790 Supplier ACROBiosystems Supplier No. ILBCM52W81MG
Recombinant Protein;1MG;Cynomolgus IL-23A & Mouse IL-12B Heterodimer Protein, His Tag&Tag Free (ILB-CM52W8) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Pro 189 (IL23A) & Met 23 - Ser 335 (IL12B) (Accession # A0A2K5TLV4-1 (IL23A) & P43432-1 (IL12B)).;Cynomolgus IL-23A & Mouse IL-12B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of IL-23 alpha and IL-12 beta, has a calculated MW of 20.3 kDa (IL-23 alpha) and 35.8 kDa (IL-12 beta). Subunit IL-23 alpha is fused with a polyhistidine tag at the N-terminus and subunit IL-12 beta contains no tag. The reducing (R) protein migrates as 20-23 kDa (IL-23 alpha) and 40-42 kDa and 43-46 kDa (IL-12 beta) respectively due to glycosylation.;Immobilized Biotinylated Human IL-23 R, Fc,Avitag (Cat. No. ILR-H82F3) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Cynomolgus IL-23A & Mouse IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-CM52W8) with a li