Learn More
ACROBiosystems Cynomolgus CD24 Protein, Fc Tag
Supplier:  ACROBiosystems CD4C52571MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus CD24, Fc Tag (CD4-C5257) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Lys 54 (Accession # I7GKK1-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 29.3 kDa. The protein migrates as 42-50 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-201-9486