Learn More
ACROBiosystems HCoV-229E S1 protein, His Tag
Supplier:  ACROBiosystems SINV52H41MG
Encompass_Preferred
Recombinant Protein;1MG;HCoV-229E S1 protein, His Tag (SIN-V52H4) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Ala 536 (Accession # P15423-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 58.8 kDa. The protein migrates as 70-110 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-201-9593