Learn More
ACROBiosystems HCoV-HKU1(isolate N5) S1 protein, His Tag
Catalog No. 502019589
Encompass_Preferred
Recombinant Protein;1MG;HCoV-HKU1(isolate N5) S1 protein, His Tag (SIN-V52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 13 - Arg 756 (Accession # Q0ZME7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.6 kDa.
Catalog No. 50-201-9589 Supplier ACROBiosystems Supplier No. SINV52H61MG
Recombinant Protein;1MG;HCoV-HKU1(isolate N5) S1 protein, His Tag (SIN-V52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 13 - Arg 756 (Accession # Q0ZME7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.6 kDa.