Learn More
ACROBiosystems HCoV-NL63 S1 protein, His Tag
Catalog No. 502019595
Encompass_Preferred
Recombinant Protein;1MG;HCoV-NL63 S1 protein, His Tag (SIN-V52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 19 - Val 717 (Accession # Q6Q1S2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 79.4 kDa. The protein migrates as 95-110 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-201-9595 Supplier ACROBiosystems Supplier No. SINV52H31MG
Recombinant Protein;1MG;HCoV-NL63 S1 protein, His Tag (SIN-V52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 19 - Val 717 (Accession # Q6Q1S2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 79.4 kDa. The protein migrates as 95-110 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.