Learn More
ACROBiosystems Human IL-13 R alpha 2 Protein, Fc Tag (MALS verified)
Catalog No. 502019458
Encompass_Preferred
Recombinant Protein;1MG;Human IL-13RA2, Fc Tag (MALS verified) (IL2-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # NP_000631.1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 63.5 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13, His Tag (Cat. No. IL3-H52H4) at 5 μg/mL (100 μL/well) can bind Human IL-13RA2, Fc Tag (Cat. No. IL2-H5256) with a linear range of 5-39 ng/mL (QC tested).;Immobilized Biotinylated Human IL-13, His,Avitag (Cat. No. IL3-H82E5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human IL-13RA2, Fc Tag (Cat. No. IL2-H5256) with a linear range of 0.5-8 ng/mL (Routinely tested).
Catalog No. 50-201-9458 Supplier ACROBiosystems Supplier No. IL2H52561MG
Recombinant Protein;1MG;Human IL-13RA2, Fc Tag (MALS verified) (IL2-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # NP_000631.1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 63.5 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13, His Tag (Cat. No. IL3-H52H4) at 5 μg/mL (100 μL/well) can bind Human IL-13RA2, Fc Tag (Cat. No. IL2-H5256) with a linear range of 5-39 ng/mL (QC tested).;Immobilized Biotinylated Human IL-13, His,Avitag (Cat. No. IL3-H82E5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human IL-13RA2, Fc Tag (Cat. No. IL2-H5256) with a linear range of 0.5-8 ng/mL (Routinely tested).