Learn More
ACROBiosystems Human IL-8 / CXCL8 Protein, Fc Tag
Catalog No. 502019695
Encompass_Preferred
Recombinant Protein;1MG;Human IL-8, Fc Tag (IL8-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession # P10145-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 34.8 kDa. The protein migrates as 37-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-8, Fc Tag (Cat. No. IL8-H5258) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human IL-8 Antibody, Human IgG1 with a linear range of 0.2-4 ng/mL (QC tested).
Catalog No. 50-201-9695 Supplier ACROBiosystems Supplier No. IL8H52581MG
Recombinant Protein;1MG;Human IL-8, Fc Tag (IL8-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession # P10145-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 34.8 kDa. The protein migrates as 37-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-8, Fc Tag (Cat. No. IL8-H5258) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human IL-8 Antibody, Human IgG1 with a linear range of 0.2-4 ng/mL (QC tested).