Learn More
ACROBiosystems Human Her2 / ErbB2 (498-648) Protein, His Tag
Catalog No. 502019758
Encompass_Preferred
Recombinant Protein;1MG;Human Her2 (498-648), His Tag (HE2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 498 - Ala 648 (Accession # P04626-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.5 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Her2 (498-648), His Tag (Cat. No. HE2-H52H4) at 1 μg/mL (100 μL/well) can bind Trastuzumab with a linear range of 0.1-13 ng/mL (QC tested).
Catalog No. 50-201-9758 Supplier ACROBiosystems Supplier No. HE2H52H41MG
Recombinant Protein;1MG;Human Her2 (498-648), His Tag (HE2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asn 498 - Ala 648 (Accession # P04626-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.5 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Her2 (498-648), His Tag (Cat. No. HE2-H52H4) at 1 μg/mL (100 μL/well) can bind Trastuzumab with a linear range of 0.1-13 ng/mL (QC tested).