Learn More
ACROBiosystems Human CD73 Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems CD3H52521MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD73, Mouse IgG2a Fc Tag (CD3-H5252) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Ser 549 (Accession # P21589-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 84.9 kDa. The protein migrates as 95-100 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human CD73, Mouse IgG2a Fc Tag (Cat. No. CD3-H5252) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD22 Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-201-9488