Learn More
ACROBiosystems Human JAK1 Protein, His Tag
Supplier:  ACROBiosystems JA1H55471MG
Encompass_Preferred
Recombinant Protein;1MG;Human JAK1, His Tag (JA1-H5547) is expressed from Baculovirus-Insect cells. It contains AA Ala 561 - Asn 852 (Accession # P23458-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 35.7 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-201-9782