Learn More
ACROBiosystems Human LIGHT / TNFSF14 Protein, Fc Tag (active trimer) (MALS verified)
Catalog No. 502019806
Encompass_Preferred
Recombinant Protein;1MG;Human LIGHT, Fc Tag (active trimer) (MALS verified) (LIT-H5269) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1) trimer Design.;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 82.6 kDa. The protein migrates as 116 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Human LIGHT, Fc Tag (active trimer) (MALS verified) (Cat. No. LIT-H5269) with a linear range of 0.2-4 ng/mL (QC tested).;Human LIGHT, Fc Tag (active trimer) (MALS verified) (Cat. No. LIT-H5269) induced cytotoxicity in HT-29 cells. The ED50 for this effect is 1.29-2.38 ng/mL (Routinely tested).
Catalog No. 50-201-9806 Supplier ACROBiosystems Supplier No. LITH52691MG
Recombinant Protein;1MG;Human LIGHT, Fc Tag (active trimer) (MALS verified) (LIT-H5269) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1) trimer Design.;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 82.6 kDa. The protein migrates as 116 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Human LIGHT, Fc Tag (active trimer) (MALS verified) (Cat. No. LIT-H5269) with a linear range of 0.2-4 ng/mL (QC tested).;Human LIGHT, Fc Tag (active trimer) (MALS verified) (Cat. No. LIT-H5269) induced cytotoxicity in HT-29 cells. The ED50 for this effect is 1.29-2.38 ng/mL (Routinely tested).