Learn More
ACROBiosystems Human LAG3 / CD223 Protein, Fc Tag (HPLC-verified)
Catalog No. 502019823
Encompass_Preferred
Recombinant Protein;1MG;Human LAG-3, Fc Tag (HPLC-verified) (LA3-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.8 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Fc Tag (HPLC-verified) (Cat. No. LA3-H5255) at 2 μg/mL (100 μL/well) can bind Madarex LAG-3 MAb with a linear range of 0.4-3 ng/mL (QC tested).
Catalog No. 50-201-9823 Supplier ACROBiosystems Supplier No. LA3H52551MG
Recombinant Protein;1MG;Human LAG-3, Fc Tag (HPLC-verified) (LA3-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.8 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Fc Tag (HPLC-verified) (Cat. No. LA3-H5255) at 2 μg/mL (100 μL/well) can bind Madarex LAG-3 MAb with a linear range of 0.4-3 ng/mL (QC tested).