Learn More
ACROBiosystems Human ROR1 (308-395, Kringle domain) Protein, His Tag
Catalog No. 502019825
Encompass_Preferred
Recombinant Protein;1MG;Human ROR1 (308-395, Kringle domain), His Tag (RO1-H5223) is expressed from human 293 cells (HEK293). It contains AA Asn 308 - Asp 395 (Accession # Q01973-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.2 kDa. The protein migrates as 16-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9825 Supplier ACROBiosystems Supplier No. RO1H52231MG
Recombinant Protein;1MG;Human ROR1 (308-395, Kringle domain), His Tag (RO1-H5223) is expressed from human 293 cells (HEK293). It contains AA Asn 308 - Asp 395 (Accession # Q01973-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.2 kDa. The protein migrates as 16-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.