Learn More
ACROBiosystems Human APRIL / TNFSF13 Protein, His,Flag Tag, active trimer (MALS verified)
Supplier:  ACROBiosystems APLH52D11MG
Encompass_Preferred
Recombinant Protein;1MG;Human APRIL, His,Flag Tag, active trimer (APL-H52D1) is expressed from human 293 cells (HEK293). It contains AA Gln 111 - Leu 250 (Accession # O75888-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 50.2 kDa. The protein migrates as 50-55 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 2 μg/mL (100 μL/well) can bind Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) with a linear range of 10-39 ng/mL (QC tested).;Immobilized Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) at 1 μg/mL (100 μL/well) can bind Human TACI/TNFRSF13B, Fc Tag (Cat. No. TAI-H5256) with a linear range of 0.8-25 ng/mL (Routinely tested).
Catalog No. 50-201-9605