Learn More
ACROBiosystems Human MICA Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems MIAH52531MG
Encompass_Preferred
Recombinant Protein;1MG;Human MICA, Fc Tag (MALS verified) (MIA-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 24- Gln 308 (Accession # AAH16929.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 59.3 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human NKG2D, His Tag (Cat. No. NKD-H5245) at 2 μg/mL (100 μL/well) can bind Human MICA, Fc Tag (Cat. No. MIA-H5253) with a linear range of 0.5-2 ng/mL (QC tested).;Immobilized Human MICA, Fc Tag (Cat. No. MIA-H5253) at 1 μg/mL (100 μL/well) can bind Human NKG2D, Mouse IgG2a Fc Tag (Cat. No. NKD-H5259) with a linear range of 0.6-20 ng/mL (Routinely tested).
Catalog No. 50-201-9618