Learn More
ACROBiosystems Human Integrin alpha M beta 2 (ITGAM&ITGB2) Heterodimer Protein, His Tag&Tag Free
Catalog No. 502019655
Encompass_Preferred
Recombinant Protein;1MG;Human ITGAM&ITGB2 Heterodimer Protein, His Tag&Tag Free (IT2-H52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 17 - Asn 1105 (ITGAM) & Gln 23 - Asn 700 (ITGB2) (Accession # P11215-2 (ITGAM) & P05107-1 (ITGB2)).;Human ITGAM&ITGB2 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAM and ITGB2, has a calculated MW of 127.2 kDa (ITGAM) and 80.2 kDa (ITGB2). Subunit ITGAM is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB2 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 150-180 kDa (ITGAM) and 85-95 kDa (ITGB2) respectively due to glycosylation.
Catalog No. 50-201-9655 Supplier ACROBiosystems Supplier No. IT2H52W41MG
Recombinant Protein;1MG;Human ITGAM&ITGB2 Heterodimer Protein, His Tag&Tag Free (IT2-H52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 17 - Asn 1105 (ITGAM) & Gln 23 - Asn 700 (ITGB2) (Accession # P11215-2 (ITGAM) & P05107-1 (ITGB2)).;Human ITGAM&ITGB2 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAM and ITGB2, has a calculated MW of 127.2 kDa (ITGAM) and 80.2 kDa (ITGB2). Subunit ITGAM is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB2 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 150-180 kDa (ITGAM) and 85-95 kDa (ITGB2) respectively due to glycosylation.