Learn More
ACROBiosystems Human BAFF / TNFSF13B / CD257 Protein, His,Flag Tag, active trimer (MALS verified)
Catalog No. 502019435
Encompass_Preferred
Recombinant Protein;1MG;Human BAFF, His,Flag Tag, active trimer (BAF-H52D4) is expressed from human 293 cells (HEK293). It contains AA Glu 140 - Leu 285 (Accession # AAH20674.1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 52.5 kDa. The protein migrates as 55-60 kDa under non-reducing (NR) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human BAFFR, Fc Tag (Cat. No. BAR-H5257) at 2 μg/mL (100 μL/well) can bind Human BAFF, His,Flag Tag, active trimer (Cat. No. BAF-H52D4) with a linear range of 4-63 ng/mL when detected by Monoclonal anti FLAG (HRP) antibody (QC tested).;Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 2 μg/mL (100 μL/well) can bind Human BAFF, His,Flag Tag, active trimer (Cat. No. BAF-H52D4) with a linear range of 2-16 ng/mL when detected by Monoclonal anti FLAG (HRP) antibody (Routinely tested).;Human BAFFR Protein, Fc Tag (Cat. No. BAR-H5257) captured on CM5 chip via A
Catalog No. 50-201-9435 Supplier ACROBiosystems Supplier No. BAFH52D41MG
Recombinant Protein;1MG;Human BAFF, His,Flag Tag, active trimer (BAF-H52D4) is expressed from human 293 cells (HEK293). It contains AA Glu 140 - Leu 285 (Accession # AAH20674.1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 52.5 kDa. The protein migrates as 55-60 kDa under non-reducing (NR) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human BAFFR, Fc Tag (Cat. No. BAR-H5257) at 2 μg/mL (100 μL/well) can bind Human BAFF, His,Flag Tag, active trimer (Cat. No. BAF-H52D4) with a linear range of 4-63 ng/mL when detected by Monoclonal anti FLAG (HRP) antibody (QC tested).;Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 2 μg/mL (100 μL/well) can bind Human BAFF, His,Flag Tag, active trimer (Cat. No. BAF-H52D4) with a linear range of 2-16 ng/mL when detected by Monoclonal anti FLAG (HRP) antibody (Routinely tested).;Human BAFFR Protein, Fc Tag (Cat. No. BAR-H5257) captured on CM5 chip via A