Learn More
ACROBiosystems MERS Nucleocapsid protein, His Tag
Supplier:  ACROBiosystems NUNM52H51MG
Encompass_Preferred
Recombinant Protein;1MG;MERS Nucleocapsid protein, His Tag (NUN-M52H5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Asp 413 (Accession # K0BVN3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.7 kDa.
Catalog No. 50-201-9460