Learn More
ACROBiosystems Mouse GUCY2C / Guanylyl cyclase C Protein, His Tag
Supplier:  ACROBiosystems GUCM52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse GUCY2C, His Tag (GUC-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Met 433 (Accession # Q3UWA6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 60-100 kD under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9638