Learn More
ACROBiosystems Mouse Mesothelin / MSLN (298-600) Protein, His Tag (MALS verified)
Catalog No. 502019687
Encompass_Preferred
Recombinant Protein;1MG;Mouse Mesothelin (298-600), His Tag (MSN-M52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 42-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Mesothelin (298-600), His Tag (Cat. No. MSN-M52H3) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CA125, Fc,Avitag (Cat. No. CA5-H82F4) with a linear range of 0.6-10 ng/mL (QC tested).
Catalog No. 50-201-9687 Supplier ACROBiosystems Supplier No. MSNM52H31MG
Recombinant Protein;1MG;Mouse Mesothelin (298-600), His Tag (MSN-M52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 42-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Mesothelin (298-600), His Tag (Cat. No. MSN-M52H3) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CA125, Fc,Avitag (Cat. No. CA5-H82F4) with a linear range of 0.6-10 ng/mL (QC tested).