Learn More
ACROBiosystems Mouse FGL1 Protein, Fc Tag
Supplier:  ACROBiosystems FG1M52581MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse FGL1, Fc Tag (FG1-M5258) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 314 (Accession # Q71KU9-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 60.5 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse LAG-3, His Tag (Cat. No. LA3-M52H5) at 5 μg/mL (100 μL/well) can bind Mouse FGL1, Fc Tag (Cat. No. FG1-M5258) with a linear range of 0.008-0.5 μg/mL (QC tested).
Catalog No. 50-201-9713