Learn More
ACROBiosystems Mouse IL-17F / IL-24 Protein, His Tag
Catalog No. 502019776
Encompass_Preferred
Recombinant Protein;1MG;Mouse IL-17F, His Tag (ILF-M5249) is expressed from human 293 cells (HEK293). It contains AA Arg 29 - Ala 161 (Accession # Q7TNI7-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 24-25 kDa and 27-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Mouse IL-17F, His Tag (Cat. No. ILF-M5249) with an affinity constant of 2.99 μM as determined in a SPR assay (Biacore 8K) (Routinely tested).
Catalog No. 50-201-9776 Supplier ACROBiosystems Supplier No. ILFM52491MG
Recombinant Protein;1MG;Mouse IL-17F, His Tag (ILF-M5249) is expressed from human 293 cells (HEK293). It contains AA Arg 29 - Ala 161 (Accession # Q7TNI7-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 24-25 kDa and 27-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Mouse IL-17F, His Tag (Cat. No. ILF-M5249) with an affinity constant of 2.99 μM as determined in a SPR assay (Biacore 8K) (Routinely tested).